Imidazo[1,2-a]pyridine-7-carboxylic acid hydrochloride

Lieferant: Apollo Scientific

Synonyme: imidazo[1,2-a]pyridine-7-carboxylic acid hydrochloride

OR111503-1G
APOSOR111503-1GEA 0 EUR
APOSOR111503-1G
Imidazo[1,2-a]pyridine-7-carboxylic acid hydrochloride
Imidazo[1,2-a]pyridine-7-carboxylic acid hydrochloride
Formel: C₈H₆N₂O₂·ClH
Molecular Weight: 198,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09954929
CAS-Nummer: 1423031-35-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR