2-Bromonaphthalen-1-ol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-1-naphthalenol

OR111504-1G
APOSOR111504-1GEA 21.9 EUR
APOSOR111504-1G
2-Bromonaphthalen-1-ol
2-Bromonaphthalen-1-ol
Formel: C₁₀H₇BrO
Molecular Weight: 223.07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00274158
CAS-Nummer: 771-15-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR