2',6'-Dimethylacetanilid

Lieferant: Apollo Scientific

Synonyme: N-(2,6-Dimethylphenyl)acetamid, Ethansäure(2,6-dimethylphenyl)amid

OR111521-5G
APOSOR111521-5GEA 21.9 EUR
APOSOR111521-5G
2',6'-Dimethylacetanilid
2',6'-Dimethylacetanilid
Formel: C₁₀H₁₃NO
Molecular Weight: 163,22 g/mol
Schmelzpunkt: 182…184 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008675
CAS-Nummer: 2198-53-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR