6-Chloroimidazo[1,2-a]pyridine-3-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR111524-1G
APOSOR111524-1GEA 440 EUR
APOSOR111524-1G
6-Chloroimidazo[1,2-a]pyridine-3-carbohydrazide
6-Chloroimidazo[1,2-a]pyridine-3-carbohydrazide
Formel: C₈H₇ClN₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD06496218
CAS-Nummer: 900018-73-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR