Methyl 3-amino-2,6-dibromo-4-methylbenzoate
Lieferant: Apollo Scientific
|
Achtung
|
OR111526-1G
APOSOR111526-1GEA
339
EUR
APOSOR111526-1G
Methyl 3-amino-2,6-dibromo-4-methylbenzoate
Methyl 3-amino-2,6-dibromo-4-methylbenzoate
Formel:
C₉H₉Br₂NO₂ Molecular Weight: 322.981 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20923513 CAS-Nummer: 1352397-65-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...