3,6-Dichloroimidazo[1,2-a]pyridine-2-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR111529-1G
APOSOR111529-1GEA 24.8 EUR
APOSOR111529-1G
3,6-Dichloroimidazo[1,2-a]pyridine-2-carbohydrazide
3,6-Dichloroimidazo[1,2-a]pyridine-2-carbohydrazide
Formel: C₈H₆Cl₂N₄O
Molecular Weight: 245,065 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31629938
CAS-Nummer: 2244083-76-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR