4,6-Dihydroxynicotinohydrazide

Lieferant: Apollo Scientific

OR111534-1G
APOSOR111534-1GEA 0 EUR
APOSOR111534-1G
4,6-Dihydroxynicotinohydrazide
4,6-Dihydroxynicotinohydrazide
Formel: C₆H₇N₃O₃
Molecular Weight: 169,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12827897
CAS-Nummer: 5466-46-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR