3-Bromoimidazo[1,2-a]pyridine-6-carbohydrazide
Lieferant: Apollo Scientific
|
Achtung
|
OR111535-1G
APOSOR111535-1GEA
174
EUR
APOSOR111535-1G
3-Bromoimidazo[1,2-a]pyridine-6-carbohydrazide
3-Bromoimidazo[1,2-a]pyridine-6-carbohydrazide
Formel:
C₈H₇BrN₄O Lagertemperatur: Raumtemperatur |
MDL:
MFCD12827905 CAS-Nummer: 1215799-36-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...