Ethyl 2-(3-nitrophenyl)pyridine-3-carboxylate

Lieferant: Apollo Scientific

OR111536-1G
APOSOR111536-1GEA 226 EUR
APOSOR111536-1G
Ethyl 2-(3-nitrophenyl)pyridine-3-carboxylate
Ethyl 2-(3-nitrophenyl)pyridine-3-carboxylate
Formel: C₁₄H₁₂N₂O₄
Molecular Weight: 272,256 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31629941
CAS-Nummer: 2244083-82-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR