2-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine
Lieferant: Apollo Scientific
|
Achtung
|
OR111537-1G
APOSOR111537-1GEA
935
EUR
APOSOR111537-1G
2-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine
2-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine
Formel:
C₈H₁₀N₂O Molecular Weight: 150,178 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18427849 CAS-Nummer: 26323-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...