Ethyl 2-acetyl-2H-indazole-5-carboxylate

Lieferant: Apollo Scientific
Achtung

OR111538-1G
APOSOR111538-1GEA 58 EUR
APOSOR111538-1G
Ethyl 2-acetyl-2H-indazole-5-carboxylate
Ethyl 2-acetyl-2H-indazole-5-carboxylate
Formel: C₁₂H₁₂N₂O₃
Molecular Weight: 232,235 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29761529
CAS-Nummer: 2089300-87-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR