4-Methyl-5-nitroimidazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 1H-Imidazole, 4-methyl-5-nitro-, 5-methyl-4-nitro-1H-imidazole, 4-METHYL-5-NITRO-1H-IMIDAZOLE

OR111542-1G OR111542-25G OR111542-5G
APOSOR111542-1GEA 21.9 EUR
APOSOR111542-1G APOSOR111542-25G APOSOR111542-5G
4-Methyl-5-nitroimidazole
4-Methyl-5-nitroimidazole
Formel: C₄H₅N₃O₂
Molecular Weight: 127.103 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01721395
CAS-Nummer: 14003-66-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR