2,5-Dichloro-4-nitro-1H-imidazole

Lieferant: Apollo Scientific
Achtung

OR111543-1G
APOSOR111543-1GEA 310 EUR
APOSOR111543-1G
2,5-Dichloro-4-nitro-1H-imidazole
2,5-Dichloro-4-nitro-1H-imidazole
Formel: C₃HCl₂N₃O₂
Molecular Weight: 181,965 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00748952
CAS-Nummer: 15965-32-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR