tert-Butyl-4-[(5-aminopyridin-2-yl)oxy]piperidine-1-carboxylate
Lieferant: Apollo Scientific
OR111547-1G
APOSOR111547-1GEA
0
EUR
APOSOR111547-1G
tert-Butyl-4-[(5-aminopyridin-2-yl)oxy]piperidine-1-carboxylate
tert-Butyl-4-[(5-aminopyridin-2-yl)oxy]piperidine-1-carboxylate
Formel:
C₁₅H₂₃N₃O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD12026334 CAS-Nummer: 346665-41-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...