1-(4'-Pentyl-1,1'-biphenyl-4-yl)ethanol

Lieferant: Apollo Scientific

OR111548-1G
APOSOR111548-1GEA 0 EUR
APOSOR111548-1G
1-(4'-Pentyl-1,1'-biphenyl-4-yl)ethanol
1-(4'-Pentyl-1,1'-biphenyl-4-yl)ethanol
Formel: C₁₉H₂₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD18089273
CAS-Nummer: 117553-23-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR