[1,2,4]Triazolo[4,3-b]pyridazin-6-amine

Lieferant: Apollo Scientific
Achtung

OR111549-1G
APOSOR111549-1GEA 323 EUR
APOSOR111549-1G
[1,2,4]Triazolo[4,3-b]pyridazin-6-amine
[1,2,4]Triazolo[4,3-b]pyridazin-6-amine
Formel: C₅H₅N₅
Lagertemperatur: Raumtemperatur
MDL: MFCD09991762
CAS-Nummer: 19195-46-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR