3-Amino-3-(3-bromo-4-hydroxyphenyl)propanoic acid

Lieferant: Apollo Scientific
Achtung

OR111557-1G
APOSOR111557-1GEA 395 EUR
APOSOR111557-1G
3-Amino-3-(3-bromo-4-hydroxyphenyl)propanoic acid
3-Amino-3-(3-bromo-4-hydroxyphenyl)propanoic acid
Formel: C₉H₁₀BrNO₃
Molecular Weight: 260,085 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03002522
CAS-Nummer: 682804-40-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR