Methyl 3-iodo-4-nitro-1H-indazole-6-carboxylate

Lieferant: Apollo Scientific
Achtung

OR111562-250MG
APOSOR111562-250MGEA 499 EUR
APOSOR111562-250MG
Methyl 3-iodo-4-nitro-1H-indazole-6-carboxylate
Methyl 3-iodo-4-nitro-1H-indazole-6-carboxylate
Formel: C₉H₆IN₃O₄
Molecular Weight: 347,066 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08272206
CAS-Nummer: 885520-73-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR