3-Iodo-4-nitro-1H-indazole-6-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR111563-250MG
APOSOR111563-250MGEA
174
EUR
APOSOR111563-250MG
3-Iodo-4-nitro-1H-indazole-6-carboxylic acid
3-Iodo-4-nitro-1H-indazole-6-carboxylic acid
Formel:
C₈H₄IN₃O₄ Molecular Weight: 333,04 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07781727 CAS-Nummer: 885521-14-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...