Benzofuran-5-carbonitrile

Lieferant: Apollo Scientific
Gefahr

OR111564-1G OR111564-5G
APOSOR111564-1GEA 94.4 EUR
APOSOR111564-1G APOSOR111564-5G
Benzofuran-5-carbonitrile
Benzofuran-5-carbonitrile
Formel: C₉H₅NO
Lagertemperatur: Raumtemperatur
MDL: MFCD05664402
CAS-Nummer: 79002-39-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR