Ethyl 2-methylimidazo[1,2-a]pyrimidine-3-carboxylate
Lieferant: Apollo Scientific
OR111570-1G
APOSOR111570-1GEA
0
EUR
APOSOR111570-1G
Ethyl 2-methylimidazo[1,2-a]pyrimidine-3-carboxylate
Ethyl 2-methylimidazo[1,2-a]pyrimidine-3-carboxylate
Formel:
C₁₀H₁₁N₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD05997359 CAS-Nummer: 62772-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...