2-Methylimidazo[1,2-a]pyrimidine-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR111571-1G
APOSOR111571-1GEA 266 EUR
APOSOR111571-1G
2-Methylimidazo[1,2-a]pyrimidine-3-carboxylic acid
2-Methylimidazo[1,2-a]pyrimidine-3-carboxylic acid
Formel: C₈H₇N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD07368659
CAS-Nummer: 90830-11-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR