(4-Chloro-2-cyclohexylphenoxy)acetic acid
Lieferant: Apollo Scientific
OR111586-1G
APOSOR111586-1GEA
310
EUR
APOSOR111586-1G
(4-Chloro-2-cyclohexylphenoxy)acetic acid
(4-Chloro-2-cyclohexylphenoxy)acetic acid
Formel:
C₁₄H₁₇ClO₃ Molecular Weight: 268,736 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09048751 CAS-Nummer: 19774-97-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...