1-(4-Chlorophenyl)ethanone (2,4,6-trichlorophenyl)hydrazone

Lieferant: Apollo Scientific

OR111588-1G
APOSOR111588-1GEA 493 EUR
APOSOR111588-1G
1-(4-Chlorophenyl)ethanone (2,4,6-trichlorophenyl)hydrazone
1-(4-Chlorophenyl)ethanone (2,4,6-trichlorophenyl)hydrazone
Formel: C₁₄H₁₀Cl₄N₂
Molecular Weight: 348,055 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01163929
CAS-Nummer: 394664-62-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR