1-Thien-2-ylethanone (2,4,6-trichlorophenyl)hydrazone

Lieferant: Apollo Scientific

OR111590-1G
APOSOR111590-1GEA 493 EUR
APOSOR111590-1G
1-Thien-2-ylethanone (2,4,6-trichlorophenyl)hydrazone
1-Thien-2-ylethanone (2,4,6-trichlorophenyl)hydrazone
Formel: C₁₂H₉Cl₃N₂S
Molecular Weight: 319,637 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01163940
CAS-Nummer: 194923-67-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR