2-(2-Bromo-5-methoxyphenyl)-1,3,4-oxadiazole

Lieferant: Apollo Scientific

OR111591-1G
APOSOR111591-1GEA 310 EUR
APOSOR111591-1G
2-(2-Bromo-5-methoxyphenyl)-1,3,4-oxadiazole
2-(2-Bromo-5-methoxyphenyl)-1,3,4-oxadiazole
Formel: C₉H₇BrN₂O₂
Molecular Weight: 255,068 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09945508
CAS-Nummer: 1016843-27-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR