Ethyl 5-bromo-3-methyl-1-benzofuran-2-carboxylate

Lieferant: Apollo Scientific

OR111595-1G
APOSOR111595-1GEA 198 EUR
APOSOR111595-1G
Ethyl 5-bromo-3-methyl-1-benzofuran-2-carboxylate
Ethyl 5-bromo-3-methyl-1-benzofuran-2-carboxylate
Formel: C₁₂H₁₁BrO₃
Molecular Weight: 283,118 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03965818
CAS-Nummer: 150612-59-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR