5-Bromo-3-methyl-1-benzofuran

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-3-methylbenzofuran

OR111596-1G
APOSOR111596-1GEA 190 EUR
APOSOR111596-1G
5-Bromo-3-methyl-1-benzofuran
5-Bromo-3-methyl-1-benzofuran
Formel: C₉H₇BrO
Molecular Weight: 211,055 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09836519
CAS-Nummer: 33118-85-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR