Ethyl (2-acetyl-5-bromophenoxy)acetate

Lieferant: Apollo Scientific

OR111597-1G
APOSOR111597-1GEA 223 EUR
APOSOR111597-1G
Ethyl (2-acetyl-5-bromophenoxy)acetate
Ethyl (2-acetyl-5-bromophenoxy)acetate
Formel: C₁₂H₁₃BrO₄
Molecular Weight: 301,133 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27932676
CAS-Nummer: 91721-00-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR