(2-Acetyl-5-bromophenoxy)acetic acid
Lieferant: Apollo Scientific
OR111598-1G
APOSOR111598-1GEA
248
EUR
APOSOR111598-1G
(2-Acetyl-5-bromophenoxy)acetic acid
(2-Acetyl-5-bromophenoxy)acetic acid
Formel:
C₁₀H₉BrO₄ Molecular Weight: 273,08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04446107 CAS-Nummer: 90772-69-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...