(2-Acetyl-4-chlorophenoxy)acetic acid

Lieferant: Apollo Scientific

OR111600-1G
APOSOR111600-1GEA 371 EUR
APOSOR111600-1G
(2-Acetyl-4-chlorophenoxy)acetic acid
(2-Acetyl-4-chlorophenoxy)acetic acid
Formel: C₁₀H₉ClO₄
Molecular Weight: 228,629 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27947477
CAS-Nummer: 21758-94-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR