Ethyl (2-acetyl-5-chlorophenoxy)acetate

Lieferant: Apollo Scientific

OR111601-1G
APOSOR111601-1GEA 198 EUR
APOSOR111601-1G
Ethyl (2-acetyl-5-chlorophenoxy)acetate
Ethyl (2-acetyl-5-chlorophenoxy)acetate
Formel: C₁₂H₁₃ClO₄
Molecular Weight: 256,682 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27949053
CAS-Nummer: 30335-94-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR