Ethyl (2,4-dichloro-3,5-dimethylphenoxy)acetate

Lieferant: Apollo Scientific

OR111603-1G
APOSOR111603-1GEA 248 EUR
APOSOR111603-1G
Ethyl (2,4-dichloro-3,5-dimethylphenoxy)acetate
Ethyl (2,4-dichloro-3,5-dimethylphenoxy)acetate
Formel: C₁₂H₁₄Cl₂O₃
Molecular Weight: 277,144 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32069497
CAS-Nummer: 2415751-85-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR