4-Hydroxy-2-(methylsulphanyl)-6-phenyl-5-pyrimidinecarbonitrile

Lieferant: Apollo Scientific

OR111606-1G
APOSOR111606-1GEA 371 EUR
APOSOR111606-1G
4-Hydroxy-2-(methylsulphanyl)-6-phenyl-5-pyrimidinecarbonitrile
4-Hydroxy-2-(methylsulphanyl)-6-phenyl-5-pyrimidinecarbonitrile
Formel: C₁₂H₉N₃OS
Molecular Weight: 243,284 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00974848
CAS-Nummer: 97693-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR