Dimethyl 3-(chlorosulphonyl)phthalate

Lieferant: Apollo Scientific

OR111607-1G
APOSOR111607-1GEA 346 EUR
APOSOR111607-1G
Dimethyl 3-(chlorosulphonyl)phthalate
Dimethyl 3-(chlorosulphonyl)phthalate
Formel: C₁₀H₉ClO₆S
Molecular Weight: 292,693 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31621195
CAS-Nummer: 142576-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR