2-(2-Chloro-3,4-dimethoxybenzyl)-1H-isoindole-1,3(2H)-dione

Lieferant: Apollo Scientific

OR111608-1G
APOSOR111608-1GEA 346 EUR
APOSOR111608-1G
2-(2-Chloro-3,4-dimethoxybenzyl)-1H-isoindole-1,3(2H)-dione
2-(2-Chloro-3,4-dimethoxybenzyl)-1H-isoindole-1,3(2H)-dione
Formel: C₁₇H₁₄ClNO₄
Molecular Weight: 331,75 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31621196
CAS-Nummer: 2227107-24-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR