1-(4-Iodobenzyl)-1H-1,2,4-triazole

Lieferant: Apollo Scientific

OR111610-1G
APOSOR111610-1GEA 173 EUR
APOSOR111610-1G
1-(4-Iodobenzyl)-1H-1,2,4-triazole
1-(4-Iodobenzyl)-1H-1,2,4-triazole
Formel: C₉H₈IN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08060477
CAS-Nummer: 864068-93-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR