N-(2,4,6-Trimethoxyphenyl)indane-5-sulphonamide

Lieferant: Apollo Scientific

OR111612-1G
APOSOR111612-1GEA 493 EUR
APOSOR111612-1G
N-(2,4,6-Trimethoxyphenyl)indane-5-sulphonamide
N-(2,4,6-Trimethoxyphenyl)indane-5-sulphonamide
Formel: C₁₈H₂₁NO₅S
Lagertemperatur: Raumtemperatur
MDL: MFCD31621197
CAS-Nummer: 2232877-40-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR