N-(6-Bromo-8-chlorodibenzo[b,d]furan-4-yl)-N-cyclopropylamine
Lieferant: Apollo Scientific
OR111614-1G
APOSOR111614-1GEA
445
EUR
APOSOR111614-1G
N-(6-Bromo-8-chlorodibenzo[b,d]furan-4-yl)-N-cyclopropylamine
N-(6-Bromo-8-chlorodibenzo[b,d]furan-4-yl)-N-cyclopropylamine
Formel:
C₁₅H₁₁BrClNO Molecular Weight: 336,611 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31707701 CAS-Nummer: 2251053-08-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...