N-(5-Bromo-4-methyl-1,3-thiazol-2-yl)acetamide

Lieferant: Apollo Scientific

OR111616-1G
APOSOR111616-1GEA 248 EUR
APOSOR111616-1G
N-(5-Bromo-4-methyl-1,3-thiazol-2-yl)acetamide
N-(5-Bromo-4-methyl-1,3-thiazol-2-yl)acetamide
Formel: C₆H₇BrN₂OS
Lagertemperatur: Raumtemperatur
MDL: MFCD01924770
CAS-Nummer: 21478-95-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR