1-(Dibenzylamino)propan-2-one
Lieferant: Apollo Scientific
OR111617-1G
APOSOR111617-1GEA
0
EUR
APOSOR111617-1G
1-(Dibenzylamino)propan-2-one
1-(Dibenzylamino)propan-2-one
Formel:
C₁₇H₁₉NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD15480865 CAS-Nummer: 68543-42-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...