2-Quinoxalinol 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR11198-500G
APOSOR11198-500GEA
82.8
EUR
APOSOR11198-500G
2-Quinoxalinol 98%
2-Quinoxalinol
Formel:
C₈H₆N₂O Schmelzpunkt: 271…272 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006722 CAS-Nummer: 1196-57-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...