3-Amino-4-(methoxycarbonyl)benzoesäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-4-methoxycarbonylbenzoic Acid, 3-Amino-4-carbomethoxybenzoic Acid, 2-Aminoterephthalic Acid 1-Methyl Ester

OR11206-25G
APOSOR11206-25GEA 377 EUR
APOSOR11206-25G
3-Amino-4-(methoxycarbonyl)benzoesäure
1-Methyl-2-aminoterephthalate
Formel: C₉H₉NO₄
Molecular Weight: 195.17 g/mol
Schmelzpunkt: 218 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00189374
CAS-Nummer: 60728-41-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR