5-Chloro-2-methylbenzimidazole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Chloro-2-methyl-1H-benzo[d]imidazole
OR11210-1G
APOSOR11210-1GEA
21.9
EUR
APOSOR11210-1G
5-Chloro-2-methylbenzimidazole
5-Chloro-2-methylbenzimidazole
Formel:
C₈H₇ClN₂ Molecular Weight: 166.61 g/mol Schmelzpunkt: 208 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014559 CAS-Nummer: 2818-69-1 EINECS: 220-575-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...