2-Methyl-5-methoxybenzimidazol

Lieferant: Apollo Scientific
Achtung

OR11211-1G
APOSOR11211-1GEA 134 EUR
APOSOR11211-1G
2-Methyl-5-methoxybenzimidazol
2-Methyl-5-methoxybenzimidazol
Formel: C₉H₁₀N₂O
Lagertemperatur: Raumtemperatur
CAS-Nummer: 4887-81-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR