(1-Naphthoxy)essigsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Naphthalen-1-yloxy)acetic acid

OR11263-100G OR11263-25G
APOSOR11263-100GEA 550 EUR
APOSOR11263-100G APOSOR11263-25G
(1-Naphthoxy)essigsäure 98%
(1-Naphthoxy)essigsäure
Formel: C₁₂H₁₀O₃
Molecular Weight: 202.21 g/mol
Schmelzpunkt: 193…197 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003927
CAS-Nummer: 2976-75-2
EINECS: 221-023-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR