ω-Brom-4-hydroxyacetophenon 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Brom-4'-hydroxyacetophenon, 2-Brom-1-(4-hydroxyphenyl)ethan-1-on, α-Brom-4-hydroxyacetophenon, 4-Hydroxyphenacylbromid
OR11266-25G
APOSOR11266-25GEA
24.8
EUR
APOSOR11266-25G
ω-Brom-4-hydroxyacetophenon 97%
ω-Brom-4-hydroxyacetophenon
Formel:
C₈H₇BrO₂ Molecular Weight: 215,05 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00072424 CAS-Nummer: 2491-38-5 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...