2-Amino-4,6-dihydroxypyrimidin 95%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-6-hydroxy-1H-pyrimidin-4-on

OR11291-500G
APOSOR11291-500GEA 88.6 EUR
APOSOR11291-500G
2-Amino-4,6-dihydroxypyrimidin 95%
2-Amino-4,6-dihydroxypyrimidine
Formel: C₄H₅N₃O₂
Molecular Weight: 127.1 g/mol
Siedepunkt: 394
Schmelzpunkt: 300
Lagertemperatur: Raumtemperatur
MDL: MFCD00006094
CAS-Nummer: 56-09-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR