(2-Bromphenyl)essigsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2-Bromophenyl)acetic acid

OR11296-500G
APOSOR11296-500GEA 243 EUR
APOSOR11296-500G
(2-Bromphenyl)essigsäure 98%
(2-Bromphenyl)essigsäure
Formel: (C₆H₄Br)CH₂COOH
Molecular Weight: 215,05 g/mol
Schmelzpunkt: 104…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004314
CAS-Nummer: 18698-97-0
EINECS: 242-509-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR