2-Amino-6-brombenzothiazol 98%

Lieferant: Apollo Scientific
Achtung

OR11314-100G
APOSOR11314-100GEA 162 EUR
APOSOR11314-100G
2-Amino-6-brombenzothiazol 98%
2-Amino-6-bromobenzothiazole
Formel: C₇H₅BrN₂S
Molecular Weight: 229.1 g/mol
Schmelzpunkt: 213…217 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00152229
CAS-Nummer: 15864-32-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR